siRNA Details
Gene Symbol: VASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003370.3 | NP_003361.1 | 12, 13 | 1445 | ||
XM_005259199.1 | XP_005259256.1 | 12, 13 | 1430 | |||
XM_005259200.1 | XP_005259257.1 | 12, 13 | 1430 | |||
XM_017027200.1 | XP_016882689.1 | 12, 13 | 1427 | |||
GenBank mRNA | AK314812.1 | - | 12, 13 | 1412 | ||
BC026019.1 | - | 12, 13 | 1382 | 3345963 | ||
BC038224.1 | - | 12, 13 | 1363 | 5210128 | ||
KU178381.1 | - | 11, 12 | 1103 | |||
KU178382.1 | - | 12, 13 | 947 | |||
KU178383.1 | - | 12, 13 | 944 | |||
KU178384.1 | - | 11, 12 | 827 | |||
KU178385.1 | - | 9, 10 | 652 | |||
Z46389.1 | - | 12, 13 | 1357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
