siRNA Details
Gene Symbol: VASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003370.3 | NP_003361.1 | 13 | 1591 | ||
XM_005259199.1 | XP_005259256.1 | 13 | 1576 | |||
XM_005259200.1 | XP_005259257.1 | 13 | 1576 | |||
XM_017027200.1 | XP_016882689.1 | 13 | 1573 | |||
GenBank mRNA | BC026019.1 | - | 13 | 1528 | 3345963 | |
BC038224.1 | - | 13 | 1509 | 5210128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
