siRNA Details
Gene Symbol: ROPN1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012337.2 | NP_001012337.1 | 6 | 941 | ||
NM_001308313.1 | NP_001295242.1 | 7 | 987 | |||
XM_005247138.3 | XP_005247195.1 | 5 | 796 | |||
XM_006713513.3 | XP_006713576.1 | 8 | 1083 | |||
GenBank mRNA | AF231410.1 | - | 6 | 900 | ||
BC015413.1 | - | 4 | 681 | 4394326 | ||
BC141849.1 | - | 7 | 934 | 8860347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
