siRNA Details
Gene Symbol: ROPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317774.1 | NP_001304703.1 | 6 | 1004 | ||
NM_001317775.1 | NP_001304704.1 | 5 | 876 | |||
NM_017578.3 | NP_060048.2 | 7 | 1084 | |||
XM_011512933.2 | XP_011511235.1 | 7 | 1048 | |||
XM_011512934.2 | XP_011511236.1 | 7 | 1159 | |||
GenBank mRNA | AF303889.2 | - | 7 | 1084 | ||
BG219480.1 | - | 2 | 352 |
Gene Symbol: ROPN1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012337.2 | NP_001012337.1 | 6 | 1010 | ||
NM_001308313.1 | NP_001295242.1 | 7 | 1056 | |||
XM_005247138.3 | XP_005247195.1 | 5 | 865 | |||
XM_006713513.3 | XP_006713576.1 | 8 | 1152 | |||
GenBank mRNA | AF231410.1 | - | 6 | 969 | ||
BC015413.1 | - | 4 | 750 | 4394326 | ||
BC141849.1 | - | 7 | 1003 | 8860347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
