siRNA Details
Gene Symbol: KRT6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173086.4 | NP_775109.2 | 9 | 2082 | ||
GenBank mRNA | BC110639.1 | - | 9 | 2082 | 4754834 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
