siRNA Details
Gene Symbol: XPO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015024.4 | NP_055839.3 | 2 | 157 | ||
XM_017013244.1 | XP_016868733.1 | 2 | 384 | |||
XM_017013245.1 | XP_016868734.1 | 2 | 151 | |||
XM_017013246.1 | XP_016868735.1 | 2 | 380 | |||
XM_017013247.1 | XP_016868736.1 | 2 | 544 | |||
GenBank mRNA | AB018288.1 | - | 2 | 67 | ||
AF064729.1 | - | 2 | 67 | |||
AK291472.1 | - | 2 | 72 | |||
AK311616.1 | - | 2 | 250 | |||
AL597247.1 | - | 2 | 113 | |||
BC030785.1 | - | 2 | 156 | 5269230 | ||
DB460798.1 | - | 2 | 159 | |||
DC332379.1 | - | 2 | 79 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
