siRNA Details
Gene Symbol: XPO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015024.4 | NP_055839.3 | 10 | 1074 | ||
XM_017013244.1 | XP_016868733.1 | 10 | 1328 | |||
XM_017013245.1 | XP_016868734.1 | 10 | 1095 | |||
XM_017013246.1 | XP_016868735.1 | 10 | 1297 | |||
XM_017013247.1 | XP_016868736.1 | 10 | 1488 | |||
GenBank mRNA | AF064729.1 | - | 10 | 984 | ||
AK291472.1 | - | 10 | 989 | |||
AK299848.1 | - | 6 | 510 | |||
BC030785.1 | - | 10 | 1073 | 5269230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
