siRNA Details
Gene Symbol: H3F3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005324.4 | NP_005315.1 | 4 | 975 | ||
GenBank mRNA | AF218029.1 | - | 4 | 799 | ||
AK130772.1 | - | 4 | 818 | |||
BC001124.2 | - | 4 | 803 | 2988279 | ||
BC006497.2 | - | 4 | 803 | 2959756 | ||
BC012813.2 | - | 4 | 803 | 2959756 | ||
BC017558.1 | - | 4 | 807 | 3910031 | ||
BC020466.1 | - | 2 | 511 | |||
BC108701.1 | - | 4 | 782 | 6144255 | ||
BX537379.1 | - | 4 | 820 | |||
BX647280.1 | - | 4 | 818 |
Gene Symbol: H3F3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013699.2 | NP_001013721.2 | 1 | 755 | ||
GenBank mRNA | BC066906.1 | - | 1 | 755 | 5267388 | |
HQ873957.1 | - | 1 | 755 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
