siRNA Details
Gene Symbol: H3F3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005324.4 | NP_005315.1 | 4 | 1370 | ||
GenBank mRNA | AF218029.1 | - | 4 | 1194 | ||
AK054591.1 | - | 1 | 187 | |||
AK130772.1 | - | 4 | 1213 | |||
AK304607.1 | - | 4 | 1144 | |||
BX537379.1 | - | 4 | 1214 | |||
BX647280.1 | - | 4 | 1213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
