siRNA Details
Gene Symbol: E2F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083588.1 | NP_001077057.1 | 2, 3 | 380 | ||
NM_001083589.1 | NP_001077058.1 | 2, 3 | 231 | |||
NM_001951.3 | NP_001942.2 | 2, 3 | 380 | |||
GenBank mRNA | AB209185.1 | - | 2, 3 | 152 | ||
AI740797.1 | - | 2, 3 | 281 | |||
BM919591.1 | - | 2, 3 | 235 | |||
BP224536.1 | - | 2, 3 | 231 | |||
U15642.1 | - | 2, 3 | 546 | |||
U31556.1 | - | 2, 3 | 381 | |||
X86097.1 | - | 2, 3 | 373 | |||
Z78409.1 | - | 2, 3 | 377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
