siRNA Details
Gene Symbol: GPNMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005340.1 | NP_001005340.1 | 4 | 548 | ||
NM_002510.2 | NP_002501.1 | 4 | 548 | |||
XM_005249578.2 | XP_005249635.1 | 4 | 1039 | |||
XM_017011676.1 | XP_016867165.1 | 4 | 1039 | |||
XM_017011677.1 | XP_016867166.1 | 4 | 1039 | |||
XM_017011678.1 | XP_016867167.1 | 4 | 1039 | |||
GenBank mRNA | AJ505015.1 | - | 4 | 460 | ||
AK296779.1 | - | 4 | 470 | |||
AK297803.1 | - | 2 | 173 | |||
AK312279.1 | - | 4 | 477 | |||
AY359124.1 | - | 4 | 468 | |||
BC025297.1 | - | 4 | 438 | |||
BC032783.1 | - | 4 | 464 | 5177095 | ||
CD105306.1 | - | 4 | 536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
