siRNA Details
Gene Symbol: GPNMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005340.1 | NP_001005340.1 | 11 | 2216 | ||
NM_002510.2 | NP_002501.1 | 11 | 2180 | |||
XM_005249578.2 | XP_005249635.1 | 11 | 2718 | |||
XM_017011676.1 | XP_016867165.1 | 12 | 2791 | |||
XM_017011677.1 | XP_016867166.1 | 12 | 2802 | |||
XM_017011678.1 | XP_016867167.1 | 11 | 2682 | |||
GenBank mRNA | AF322909.1 | - | 11 | 2078 | ||
AK297031.1 | - | 10 | 1964 | |||
AK297051.1 | - | 11 | 1985 | |||
AK297803.1 | - | 9 | 1805 | |||
AK303071.1 | - | 9 | 1558 | |||
AY359124.1 | - | 11 | 2136 | |||
BC025297.1 | - | 11 | 2070 | |||
BC032783.1 | - | 11 | 2132 | 5177095 | ||
X76534.1 | - | 11 | 2110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
