siRNA Details
Gene Symbol: GPNMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005340.1 | NP_001005340.1 | 11 | 2308 | ||
NM_002510.2 | NP_002501.1 | 11 | 2272 | |||
XM_005249578.2 | XP_005249635.1 | 11 | 2810 | |||
XM_017011676.1 | XP_016867165.1 | 12 | 2883 | |||
XM_017011677.1 | XP_016867166.1 | 12 | 2894 | |||
XM_017011678.1 | XP_016867167.1 | 11 | 2774 | |||
GenBank mRNA | AF322909.1 | - | 11 | 2170 | ||
AK297031.1 | - | 10 | 2056 | |||
AK303071.1 | - | 9 | 1650 | |||
AY359124.1 | - | 11 | 2228 | |||
BC025297.1 | - | 11 | 2162 | |||
BC032783.1 | - | 11 | 2224 | 5177095 | ||
X76534.1 | - | 11 | 2202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
