siRNA Details
Gene Symbol: NAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004537.5 | NP_004528.1 | 15 | 3617 | ||
NM_139207.3 | NP_631946.1 | 16 | 2943 | |||
XM_006719425.3 | XP_006719488.1 | 14 | 2406 | |||
XM_011538390.2 | XP_011536692.2 | 16 | 2574 | |||
XM_011538393.2 | XP_011536695.1 | 16 | 2855 | |||
XM_017019338.1 | XP_016874827.1 | 16 | 2827 | |||
XM_017019339.1 | XP_016874828.1 | 14 | 2666 | |||
XM_017019340.1 | XP_016874829.1 | 14 | 2378 | |||
GenBank mRNA | AF086080.1 | - | 1 | 436 | ||
AF300335.2 | - | 1 | 769 | |||
AL162068.1 | - | 15 | 3256 | |||
DA137494.1 | - | 1 | 147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
