siRNA Details
Gene Symbol: NAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307924.1 | NP_001294853.1 | 2, 3 | 502 | ||
NM_004537.5 | NP_004528.1 | 2, 3 | 502 | |||
NM_139207.3 | NP_631946.1 | 2, 3 | 502 | |||
XM_011538390.2 | XP_011536692.2 | 2, 3 | 161 | |||
XM_011538393.2 | XP_011536695.1 | 2, 3 | 414 | |||
XM_017019338.1 | XP_016874827.1 | 2, 3 | 414 | |||
GenBank mRNA | AK295147.1 | - | 2, 3 | 81 | ||
AL162068.1 | - | 2, 3 | 139 | |||
BC002387.2 | - | 2, 3 | 136 | 2964631 | ||
BC015599.1 | - | 1, 2 | 351 | |||
BT007023.1 | - | 1, 2 | 15 | |||
BU183927.1 | - | 501 | ||||
DC300275.1 | - | 2, 3 | 446 | |||
DC321631.1 | - | 2, 3 | 428 | |||
M86667.1 | - | 2, 3 | 90 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
