siRNA Details
Gene Symbol: KLHL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007075.2 | NP_001007076.1 | 2 | 369 | ||
NM_015990.4 | NP_057074.3 | 1 | 431 | |||
NM_199039.3 | NP_950240.2 | 1 | 431 | |||
XM_005262655.4 | XP_005262712.1 | 2 | 403 | |||
XM_011513699.2 | XP_011512001.1 | 2 | 405 | |||
XM_011513700.2 | XP_011512002.1 | 2 | 405 | |||
XM_011513701.2 | XP_011512003.1 | 2 | 405 | |||
XM_017008273.1 | XP_016863762.1 | 2 | 405 | |||
XM_017008274.1 | XP_016863763.1 | 2 | 405 | |||
XM_017008275.1 | XP_016863764.1 | 2 | 400 | |||
XM_017008276.1 | XP_016863765.1 | 2 | 405 | |||
GenBank mRNA | AF272976.1 | - | 1 | 223 | ||
AK289785.1 | - | 1 | 431 | |||
AY172948.1 | - | 1 | 223 | |||
BC053860.1 | - | 2 | 171 | |||
BQ438207.1 | - | 193 | ||||
BX478676.1 | - | 1 | 311 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
