siRNA Details
Gene Symbol: CHD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170629.1 | NP_001164100.1 | 10 | 2385 | ||
NM_020920.3 | NP_065971.2 | 11 | 1630 | |||
GenBank mRNA | AB046784.3 | - | 10 | 1873 | ||
AK122910.1 | - | 11 | 1577 | |||
CR749315.1 | - | 11 | 1575 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
