siRNA Details
Gene Symbol: CHD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170629.1 | NP_001164100.1 | 22, 23 | 4635 | ||
NM_020920.3 | NP_065971.2 | 23, 24 | 3880 | |||
GenBank mRNA | AB046784.3 | - | 22, 23 | 4123 | ||
AK056639.1 | - | 3, 4 | 502 | |||
AK098408.1 | - | 7, 8 | 1245 | |||
AK122910.1 | - | 23, 24 | 3828 | |||
AK131077.1 | - | 8, 9 | 1459 | |||
CR749315.1 | - | 23, 24 | 3825 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
