siRNA Details
Gene Symbol: SMARCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007468.2 | NP_001007469.1 | 2 | 434 | ||
NM_001317946.1 | NP_001304875.1 | 2 | 434 | |||
NM_003073.4 | NP_003064.2 | 2 | 434 | |||
XM_011530345.2 | XP_011528647.1 | 2 | 403 | |||
GenBank mRNA | AB017523.1 | - | 2 | 195 | ||
AJ011737.1 | - | 2 | 342 | |||
AJ011738.1 | - | 2 | 342 | |||
AK021419.1 | - | 2 | 367 | |||
AK024025.1 | - | 2 | 402 | |||
AK299411.1 | - | 2 | 364 | |||
AK303620.1 | - | 2 | 385 | |||
AL389878.1 | - | 2 | 244 | |||
AU143041.1 | - | 2 | 402 | |||
AY207470.1 | - | 1 | 47 | |||
AY207471.1 | - | 1 | 47 | |||
AY207472.1 | - | 47 | ||||
BC117114.1 | - | 2 | 250 | 40125665 | ||
BC143667.1 | - | 2 | 250 | 9052181 | ||
BG705851.1 | - | 2 | 407 | |||
CR456581.1 | - | 2 | 248 | |||
HY013730.1 | - | 2 | 436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
