siRNA Details
Gene Symbol: SMARCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007468.2 | NP_001007469.1 | 9 | 1614 | ||
NM_001317946.1 | NP_001304875.1 | 9 | 1668 | |||
NM_003073.4 | NP_003064.2 | 9 | 1641 | |||
XM_011530345.2 | XP_011528647.1 | 9 | 1664 | |||
GenBank mRNA | AJ011737.1 | - | 9 | 1550 | ||
AJ011738.1 | - | 9 | 1522 | |||
AK021419.1 | - | 9 | 1547 | |||
AK024025.1 | - | 9 | 1636 | |||
AL389881.1 | - | 2 | 422 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
