siRNA Details
Gene Symbol: NUP160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318399.1 | NP_001305328.1 | 3 | 609 | ||
NM_015231.2 | NP_056046.1 | 3 | 609 | |||
NR_134636.1 | 3 | 609 | ||||
GenBank mRNA | AK026236.1 | - | 3 | 392 | ||
AK304308.1 | - | 3 | 395 | |||
AL832029.1 | - | 3 | 399 | |||
BC009822.2 | - | 3 | 570 | |||
BC125227.1 | - | 3 | 532 | 40124592 | ||
BC125228.1 | - | 3 | 532 | 40124593 | ||
BI559779.1 | - | 3 | 620 | |||
BX647634.1 | - | 3 | 394 | |||
D83781.1 | - | 3 | 378 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
