siRNA Details
Gene Symbol: EIF2AK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013703.3 | NP_001013725.2 | 9 | 1405 | ||
XM_005254392.2 | XP_005254449.1 | 9 | 1428 | |||
XM_011521599.2 | XP_011519901.1 | 9 | 1428 | |||
XM_011521600.2 | XP_011519902.1 | 9 | 1428 | |||
XM_017022219.1 | XP_016877708.1 | 9 | 1428 | |||
GenBank mRNA | AB037759.1 | - | 5 | 871 | ||
AL832907.1 | - | 4 | 749 | |||
BC078179.1 | - | 9 | 1383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
