siRNA Details
Gene Symbol: CHMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005753.2 | NP_001005753.1 | 4 | 524 | ||
NM_001193517.1 | NP_001180446.1 | 5 | 465 | |||
NM_016079.3 | NP_057163.1 | 5 | 585 | |||
NR_036454.1 | 3 | 344 | ||||
GenBank mRNA | AF151907.1 | - | 5 | 454 | ||
AF219226.1 | - | 5 | 436 | |||
AK025260.1 | - | 5 | 483 | |||
AK095433.1 | - | 2 | 2206 | |||
AK129502.1 | - | 5 | 342 | |||
AK290725.1 | - | 4 | 415 | |||
AK312353.1 | - | 5 | 531 | |||
AK315835.1 | - | 4 | 468 | |||
AY364249.1 | - | 5 | 436 | |||
BC004419.1 | - | 5 | 483 | 3660116 | ||
BI562428.1 | - | 5 | 575 | |||
DA046087.1 | - | 3 | 306 |
Gene Symbol: RNF103-CHMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198954.1 | NP_001185883.1 | 7 | 766 | ||
GenBank mRNA | AK294389.1 | - | 7 | 766 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
