siRNA Details
Gene Symbol: SUMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005781.1 | NP_001005781.1 | 6 | 526 | ||
NM_001005782.1 | NP_001005782.1 | 4 | 900 | |||
NM_003352.4 | NP_003343.1 | 5 | 975 | |||
GenBank mRNA | AB062294.1 | - | 5 | 951 | ||
BC005899.1 | - | 5 | 943 | |||
BC006462.1 | - | 5 | 959 | 3546125 | ||
BC053528.1 | - | 5 | 933 | 3452929 | ||
BC066306.1 | - | 5 | 951 | 4818792 | ||
BU532734.1 | - | 475 | ||||
U38784.1 | - | 6 | 893 | |||
U61397.1 | - | 5 | 973 | |||
U83117.1 | - | 6 | 963 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
