siRNA Details
Gene Symbol: SUMO1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002190.1 | 1 | 1167 |
Gene Symbol: SUMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005781.1 | NP_001005781.1 | 6 | 730 | ||
NM_001005782.1 | NP_001005782.1 | 4 | 1104 | |||
NM_003352.4 | NP_003343.1 | 5 | 1179 | |||
GenBank mRNA | AB062294.1 | - | 5 | 1155 | ||
BC006462.1 | - | 5 | 1163 | 3546125 | ||
BC053528.1 | - | 5 | 1137 | 3452929 | ||
BC066306.1 | - | 5 | 1155 | 4818792 | ||
BQ930026.1 | - | 1 | 34 | |||
U61397.1 | - | 5 | 1177 | |||
U83117.1 | - | 6 | 1167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
