siRNA Details
Gene Symbol: HERC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271602.1 | NP_001258531.1 | 14 | 1665 | ||
NM_014606.2 | NP_055421.1 | 15 | 1920 | |||
XM_005263327.3 | XP_005263384.1 | 16 | 2047 | |||
XM_005263329.3 | XP_005263386.1 | 15 | 1920 | |||
XM_005263330.3 | XP_005263387.1 | 15 | 1920 | |||
XM_005263331.1 | XP_005263388.1 | 15 | 1920 | |||
XM_005263332.3 | XP_005263389.1 | 15 | 1920 | |||
XM_011532397.2 | XP_011530699.1 | 2 | 160 | |||
XM_017008806.1 | XP_016864295.1 | 16 | 2047 | |||
XM_017008807.1 | XP_016864296.1 | 14 | 1643 | |||
XM_017008808.1 | XP_016864297.1 | 8 | 822 | |||
GenBank mRNA | AK289990.1 | - | 15 | 1863 | ||
AK296379.1 | - | 14 | 1641 | |||
AK296397.1 | - | 3 | 384 | |||
D25215.1 | - | 15 | 1847 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
