siRNA Details
Gene Symbol: HERC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271602.1 | NP_001258531.1 | 15, 16 | 1905 | ||
NM_014606.2 | NP_055421.1 | 16, 17 | 2160 | |||
XM_005263327.3 | XP_005263384.1 | 17, 18 | 2287 | |||
XM_005263330.3 | XP_005263387.1 | 16, 17 | 2160 | |||
XM_005263331.1 | XP_005263388.1 | 16, 17 | 2160 | |||
XM_005263332.3 | XP_005263389.1 | 16, 17 | 2160 | |||
XM_011532397.2 | XP_011530699.1 | 3, 4 | 400 | |||
XM_017008808.1 | XP_016864297.1 | 9, 10 | 1062 | |||
GenBank mRNA | AK289990.1 | - | 16, 17 | 2103 | ||
AK296379.1 | - | 15, 16 | 1881 | |||
AK296397.1 | - | 4, 5 | 624 | |||
D25215.1 | - | 16, 17 | 2087 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
