siRNA Details
Gene Symbol: MAP3K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005921.1 | NP_005912.1 | 12 | 2604 | ||
XM_017009484.1 | XP_016864973.1 | 12 | 5315 | |||
XM_017009485.1 | XP_016864974.1 | 12 | 2043 | |||
GenBank mRNA | AF042838.1 | - | 12 | 2046 | ||
AK307828.1 | - | 13 | 1824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
