siRNA Details
Gene Symbol: SPEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005876.4 | NP_005867.3 | 25 | 5304 | ||
XM_005246237.2 | XP_005246294.1 | 27 | 5032 | |||
XM_005246239.2 | XP_005246296.1 | 27 | 4847 | |||
XM_005246240.2 | XP_005246297.1 | 20 | 3049 | |||
XM_005246241.1 | XP_005246298.1 | 20 | 2943 | |||
XM_005246242.4 | XP_005246299.1 | 20 | 3068 | |||
XM_006712189.3 | XP_006712252.1 | 26 | 5037 | |||
XM_006712193.3 | XP_006712256.1 | 20 | 6168 | |||
XM_011510479.2 | XP_011508781.1 | 26 | 5470 | |||
XM_011510483.2 | XP_011508785.2 | 26 | 5067 | |||
XM_017003157.1 | XP_016858646.1 | 26 | 5067 | |||
XM_017003158.1 | XP_016858647.1 | 22 | 4643 | |||
XM_017003159.1 | XP_016858648.1 | 23 | 4826 | |||
XM_017003160.1 | XP_016858649.1 | 10 | 1239 | |||
GenBank mRNA | AB037718.1 | - | 16 | 2361 | ||
AY603755.1 | - | 24 | 4917 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
