siRNA Details
Gene Symbol: KSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014238.1 | NP_055053.1 | 9 | 1178 | ||
XM_006722154.3 | XP_006722217.1 | 8 | 1330 | |||
XM_011525429.2 | XP_011523731.1 | 8 | 1330 | |||
XM_011525430.2 | XP_011523732.1 | 8 | 1332 | |||
XM_011525431.2 | XP_011523733.1 | 8 | 1332 | |||
XM_017025267.1 | XP_016880756.1 | 8 | 1330 | |||
XM_017025268.1 | XP_016880757.1 | 8 | 1330 | |||
XM_017025269.1 | XP_016880758.1 | 8 | 1330 | |||
XM_017025270.1 | XP_016880759.1 | 7 | 1065 | |||
XM_017025271.1 | XP_016880760.1 | 9 | 2037 | |||
XM_017025272.1 | XP_016880761.1 | 9 | 2020 | |||
XM_017025273.1 | XP_016880762.1 | 8 | 1843 | |||
XM_017025274.1 | XP_016880763.1 | 8 | 1046 | |||
XM_017025275.1 | XP_016880764.1 | 8 | 1033 | |||
XM_017025276.1 | XP_016880765.1 | 9 | 1985 | |||
XM_017025277.1 | XP_016880766.1 | 4 | 728 | |||
XM_017025278.1 | XP_016880767.1 | 4 | 738 | |||
XM_017025279.1 | XP_016880768.1 | 8 | 1347 | |||
XM_017025280.1 | XP_016880769.1 | 8 | 1347 | |||
GenBank mRNA | BC035488.1 | - | 9 | 2156 | ||
BC042106.1 | - | 9 | 1177 | |||
U43586.1 | - | 5 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
