siRNA Details
Gene Symbol: DMWD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004943.1 | NP_004934.1 | 5 | 2169 | ||
GenBank mRNA | AK309497.1 | - | 2 | 637 | ||
BC019266.1 | - | 2 | 1177 | |||
L19267.1 | - | 5 | 1760 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
