siRNA Details
Gene Symbol: RPL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000994.3 | NP_000985.1 | 3 | 275 | ||
NM_001007073.1 | NP_001007074.1 | 3 | 374 | |||
NM_001007074.1 | NP_001007075.1 | 2 | 394 | |||
GenBank mRNA | AK311909.1 | - | 3 | 232 | ||
BC011514.1 | - | 1 | 201 | 2900089 | ||
BC070209.1 | - | 1 | 207 | 6738371 | ||
BF960792.1 | - | 2 | 405 | |||
BQ068496.1 | - | 312 | ||||
CF124158.1 | - | 3 | 312 | |||
X03342.1 | - | 3 | 215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
