siRNA Details
Gene Symbol: FBXO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012166.2 | NP_036298.2 | 2 | 375 | ||
XM_005251439.4 | XP_005251496.1 | 2 | 464 | |||
XM_017014618.1 | XP_016870107.1 | 2 | 581 | |||
XM_017014619.1 | XP_016870108.1 | 2 | 692 | |||
GenBank mRNA | AF174598.1 | - | 2 | 374 | ||
AF176705.1 | - | 2 | 374 | |||
AI475671.1 | - | 2 | 375 | |||
BC125125.1 | - | 2 | 454 | 40119019 | ||
BC140785.1 | - | 1 | 346 | 9021653 | ||
BC171785.1 | - | 1 | 346 | 9054439 | ||
CN411883.1 | - | 2 | 375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
