siRNA Details
Gene Symbol: FBXO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012166.2 | NP_036298.2 | 4 | 1608 | ||
XM_005251439.4 | XP_005251496.1 | 4 | 1697 | |||
XM_006716754.3 | XP_006716817.1 | 2 | 163 | |||
XM_017014618.1 | XP_016870107.1 | 4 | 1814 | |||
XM_017014619.1 | XP_016870108.1 | 4 | 1925 | |||
GenBank mRNA | AB209610.1 | - | 3 | 1196 | ||
BC125124.1 | - | 2 | 202 | 40119018 | ||
BC125125.1 | - | 4 | 1687 | 40119019 | ||
BC140785.1 | - | 3 | 1579 | 9021653 | ||
BC171785.1 | - | 3 | 1579 | 9054439 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
