siRNA Details
Gene Symbol: FBXO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012166.2 | NP_036298.2 | 2 | 443 | ||
XM_005251439.4 | XP_005251496.1 | 2 | 532 | |||
XM_017014618.1 | XP_016870107.1 | 2 | 649 | |||
XM_017014619.1 | XP_016870108.1 | 2 | 760 | |||
GenBank mRNA | AB209610.1 | - | 1 | 31 | ||
AF174598.1 | - | 2 | 442 | |||
AF176705.1 | - | 2 | 442 | |||
AI475671.1 | - | 2 | 443 | |||
BC125125.1 | - | 2 | 522 | 40119019 | ||
BC140785.1 | - | 1 | 414 | 9021653 | ||
BC171785.1 | - | 1 | 414 | 9054439 | ||
CN411883.1 | - | 2 | 443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
