siRNA Details
Gene Symbol: CABLES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031215.2 | NP_112492.2 | 7 | 973 | ||
GenBank mRNA | BC003122.1 | - | 5 | 508 | ||
BU166894.1 | - | 7 | 687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
