siRNA Details
Gene Symbol: DNAH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207437.3 | NP_997320.2 | 63, 64 | 10909 | ||
XM_005253555.3 | XP_005253612.1 | 64, 65 | 11572 | |||
XM_011538014.2 | XP_011536316.1 | 63, 64 | 11413 | |||
XM_011538015.2 | XP_011536317.1 | 63, 64 | 11383 | |||
XM_011538016.2 | XP_011536318.1 | 63, 64 | 11383 | |||
XM_011538017.2 | XP_011536319.1 | 62, 63 | 11236 | |||
XM_011538019.2 | XP_011536321.1 | 46, 47 | 8171 | |||
XM_017018960.1 | XP_016874449.1 | 62, 63 | 11287 | |||
XM_017018961.1 | XP_016874450.1 | 63, 64 | 11263 | |||
XM_017018962.1 | XP_016874451.1 | 46, 47 | 8093 | |||
GenBank mRNA | AB095937.2 | - | 38, 39 | 6624 | ||
AB290168.1 | - | 43, 44 | 7428 | |||
AK095581.1 | - | 12, 13 | 2111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
