siRNA Details
Gene Symbol: NUP205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015135.2 | NP_055950.1 | 17 | 2479 | ||
XM_005250235.3 | XP_005250292.1 | 16 | 2431 | |||
GenBank mRNA | BC136624.1 | - | 17 | 2474 | 9020614 | |
BC146784.1 | - | 17 | 2453 | 9007240 | ||
D86978.1 | - | 17 | 2453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
