siRNA Details
Gene Symbol: HIST1H2BD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138720.2 | NP_619790.1 | 2 | 733 | ||
GenBank mRNA | AJ223353.1 | - | 2 | 712 | ||
BC002842.2 | - | 2 | 709 | 3659807 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
