siRNA Details
Gene Symbol: RPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000661.4 | NP_000652.2 | 8 | 720 | ||
NM_001024921.3 | NP_001020092.1 | 8 | 878 | |||
GenBank mRNA | AB062431.1 | - | 1 | 667 | ||
AK130966.1 | - | 1 | 683 | |||
AK303908.1 | - | 2 | 624 | |||
AL832047.1 | - | 3 | 4276 | |||
AY320412.1 | - | 8 | 703 | |||
AY376242.1 | - | 7 | 837 | |||
AY597415.1 | - | 1 | 694 | |||
BC000483.2 | - | 1 | 672 | 2964733 | ||
BC004156.2 | - | 1 | 666 | 2961026 | ||
BC004206.2 | - | 1 | 666 | 2961026 | ||
BC007967.2 | - | 1 | 722 | 4304670 | ||
BC012149.1 | - | 1 | 722 | 4554838 | ||
BC031906.1 | - | 7 | 667 | 4996986 | ||
BC066318.1 | - | 1 | 687 | 5277491 | ||
BC070214.1 | - | 1 | 726 | 6605055 | ||
BC100285.1 | - | 7 | 827 | 6022858 | ||
BG829769.1 | - | 7 | 667 | |||
D14531.1 | - | 7 | 667 | |||
U09953.1 | - | 8 | 690 | |||
U21138.1 | - | 1 | 667 |
Gene Symbol: SMARCA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026573.1 | - | 12 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
