siRNA Details
Gene Symbol: TSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000368.4 | NP_000359.1 | 9, 10 | 1139 | ||
NM_001162426.1 | NP_001155898.1 | 9, 10 | 1139 | |||
NM_001162427.1 | NP_001155899.1 | 8, 9 | 986 | |||
XM_005272211.1 | XP_005272268.1 | 9, 10 | 1184 | |||
XM_006717271.1 | XP_006717334.1 | 9, 10 | 1119 | |||
XM_011518979.2 | XP_011517281.1 | 9, 10 | 1133 | |||
XM_017015096.1 | XP_016870585.1 | 10, 11 | 1598 | |||
XM_017015097.1 | XP_016870586.1 | 10, 11 | 1569 | |||
XM_017015098.1 | XP_016870587.1 | 9, 10 | 1184 | |||
XM_017015099.1 | XP_016870588.1 | 8, 9 | 1277 | |||
XM_017015100.1 | XP_016870589.1 | 8, 9 | 1248 | |||
XM_017015101.1 | XP_016870590.1 | 8, 9 | 1278 | |||
GenBank mRNA | AB190910.1 | - | 9, 10 | 1101 | ||
AF013168.1 | - | 9, 10 | 1126 | |||
AK297326.1 | - | 8, 9 | 1021 | |||
AK299654.1 | - | 7, 8 | 820 | |||
AK303030.1 | - | 8, 9 | 969 | |||
AK308412.1 | - | 9, 10 | 1122 | |||
BC047772.2 | - | 9, 10 | 1112 | |||
BC070032.1 | - | 9, 10 | 1140 | |||
BC108668.1 | - | 9, 10 | 1129 | |||
BC121000.2 | - | 9, 10 | 1096 | |||
KJ534953.1 | - | 7, 8 | 905 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
