siRNA Details
Gene Symbol: DNAH17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173628.3 | NP_775899.3 | 36 | 5730 | ||
XM_011525416.2 | XP_011523718.1 | 36 | 5766 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
