siRNA Details
Gene Symbol: HIST1H2BI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003525.2 | NP_003516.1 | 1 | 394 | ||
GenBank mRNA | BC101653.1 | - | 1 | 454 | 8069159 | |
BC101655.1 | - | 1 | 454 | 8069161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
