siRNA Details
Gene Symbol: POTEJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511683.2 | XP_011509985.1 | 15 | 3477 | ||
XM_017004741.1 | XP_016860230.1 | 16 | 3450 | |||
XM_017004742.1 | XP_016860231.1 | 10 | 2313 | |||
XM_017004743.1 | XP_016860232.1 | 8 | 2245 |
Gene Symbol: POTEE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511216.2 | XP_011509518.1 | 11 | 3192 | ||
XM_017004159.1 | XP_016859648.1 | 11 | 3223 | |||
XM_017004160.1 | XP_016859649.1 | 7 | 2191 | |||
XM_017004161.1 | XP_016859650.1 | 17 | 3905 |
Gene Symbol: POTEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099771.2 | NP_001093241.1 | 17 | 3816 | ||
XM_017004833.1 | XP_016860322.1 | 15 | 3591 | |||
XM_017004834.1 | XP_016860323.1 | 10 | 2313 | |||
XM_017004835.1 | XP_016860324.1 | 8 | 2250 | |||
GenBank mRNA | EF523384.1 | - | 17 | 3818 |
Gene Symbol: POTEI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511670.2 | XP_011509972.1 | 15 | 3588 | ||
XM_017004732.1 | XP_016860221.1 | 16 | 3567 | |||
XM_017004733.1 | XP_016860222.1 | 10 | 2313 | |||
XM_017004734.1 | XP_016860223.1 | 8 | 2250 | |||
XM_017004735.1 | XP_016860224.1 | 3 | 1210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
