siRNA Details
Gene Symbol: POTEI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277406.1 | NP_001264335.1 | 15 | 3076 | ||
XM_011511670.2 | XP_011509972.1 | 15 | 3248 | |||
XM_017004732.1 | XP_016860221.1 | 16 | 3227 | |||
XM_017004733.1 | XP_016860222.1 | 10 | 1973 | |||
XM_017004734.1 | XP_016860223.1 | 8 | 1910 | |||
XM_017004735.1 | XP_016860224.1 | 3 | 870 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
