siRNA Details
Gene Symbol: INPP5F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243195.1 | NP_001230124.1 | 3 | 459 | ||
NM_014937.3 | NP_055752.1 | 3 | 459 | |||
XM_006717720.3 | XP_006717783.1 | 3 | 2846 | |||
XM_011539524.2 | XP_011537826.1 | 3 | 222 | |||
XM_011539525.2 | XP_011537827.1 | 4 | 2929 | |||
XM_011539527.2 | XP_011537829.2 | 2 | 224 | |||
XM_011539528.2 | XP_011537830.1 | 2 | 346 | |||
XM_017015935.1 | XP_016871424.1 | 3 | 213 | |||
XM_017015936.1 | XP_016871425.1 | 6 | 847 | |||
GenBank mRNA | AB023183.1 | - | 3 | 409 | ||
AK307758.1 | - | 3 | 449 | |||
BC052367.1 | - | 3 | 344 | |||
BC067820.1 | - | 3 | 420 | |||
BC082755.1 | - | 3 | 344 | |||
BC111493.1 | - | 3 | 370 | 6166990 | ||
DB636424.1 | - | 3 | 458 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
