siRNA Details
Gene Symbol: CKAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008938.3 | NP_001008938.1 | 3 | 316 | ||
NM_014756.3 | NP_055571.2 | 3 | 316 | |||
GenBank mRNA | BC017856.1 | - | 3 | 286 | ||
BC070136.1 | - | 3 | 286 | |||
BC111043.1 | - | 3 | 268 | |||
BC120869.1 | - | 2 | 210 | 40072423 | ||
BC120870.1 | - | 2 | 208 | 40072425 | ||
BP282525.1 | - | 3 | 316 | |||
D43948.1 | - | 2 | 237 | |||
X92474.1 | - | 2 | 216 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
