siRNA Details
Gene Symbol: CKAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008938.3 | NP_001008938.1 | 21 | 2675 | ||
NM_014756.3 | NP_055571.2 | 21 | 2675 | |||
GenBank mRNA | BC120869.1 | - | 20 | 2569 | 40072423 | |
BC120870.1 | - | 20 | 2567 | 40072425 | ||
D43948.1 | - | 20 | 2596 | |||
X92474.1 | - | 20 | 2575 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
