siRNA Details
Gene Symbol: PEAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024776.3 | NP_079052.2 | 6 | 3886 | ||
XM_005254670.2 | XP_005254727.1 | 9 | 4190 | |||
XM_005254671.2 | XP_005254728.1 | 8 | 4087 | |||
XM_005254673.2 | XP_005254730.1 | 6 | 3821 | |||
XM_005254674.2 | XP_005254731.1 | 7 | 4034 | |||
XM_005254675.4 | XP_005254732.1 | 3 | 3540 | |||
XM_011522033.2 | XP_011520335.1 | 10 | 4246 | |||
XM_011522034.2 | XP_011520336.1 | 11 | 4354 | |||
XM_011522035.2 | XP_011520337.1 | 13 | 4505 | |||
XM_011522036.2 | XP_011520338.1 | 12 | 4409 | |||
XM_011522037.2 | XP_011520339.1 | 12 | 4450 | |||
XM_011522038.2 | XP_011520340.1 | 11 | 4301 | |||
XM_011522039.2 | XP_011520341.1 | 12 | 4397 | |||
XM_011522040.1 | XP_011520342.1 | 10 | 4246 | |||
XM_011522042.2 | XP_011520344.1 | 2 | 623 | |||
GenBank mRNA | AB082533.1 | - | 2 | 365 | ||
AK074157.1 | - | 2 | 1192 | |||
AK127658.1 | - | 2 | 1348 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
