siRNA Details
Gene Symbol: PEAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024776.3 | NP_079052.2 | 8 | 5782 | ||
XM_005254670.2 | XP_005254727.1 | 11 | 6086 | |||
XM_005254671.2 | XP_005254728.1 | 10 | 5983 | |||
XM_005254673.2 | XP_005254730.1 | 8 | 5717 | |||
XM_005254674.2 | XP_005254731.1 | 9 | 5930 | |||
XM_005254675.4 | XP_005254732.1 | 5 | 5436 | |||
XM_011522033.2 | XP_011520335.1 | 12 | 6142 | |||
XM_011522034.2 | XP_011520336.1 | 13 | 6250 | |||
XM_011522035.2 | XP_011520337.1 | 15 | 6401 | |||
XM_011522036.2 | XP_011520338.1 | 14 | 6305 | |||
XM_011522037.2 | XP_011520339.1 | 14 | 6346 | |||
XM_011522038.2 | XP_011520340.1 | 13 | 6197 | |||
XM_011522039.2 | XP_011520341.1 | 14 | 6293 | |||
XM_011522042.2 | XP_011520344.1 | 4 | 2519 | |||
GenBank mRNA | AB082533.1 | - | 4 | 2261 | ||
AK024793.1 | - | 1 | 1122 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
